missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzenesulfonic acid, sodium salt, technical

Product Code. 10083640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10083640 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083640 Supplier Thermo Scientific Acros Supplier No. 325910010
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25155-30-0 | C18H29NaO3S | 348.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Synonym Sodium dodecylbenzenesulfonate

Specifications

Color White to Yellow
pH 6.5 to 9.5 (1% soln.)
Infrared Spectrum Authentic
Assay Percent Range 88%
Packaging Plastic bottle
Quantity 1 kg
Formula Weight 348.48
Percent Purity ≥83%
Physical Form Crystalline Powder
Chemical Name or Material Dodecylbenzenesulfonic acid, sodium salt
compliance-icons compliance-icons
Product Identifier
  • Dodecylbenzenesulfonic acid, sodium salt, technical
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium dodecylbenzenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.