missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Dofetilide
Description
Dofetilide is a selective potassium channel blocker. Blocks KV11.1 (hERG) channels; inhibits the rapid delayed-rectifier K+ current (IKr). Displays class III antiarrhythmic properties.
Specifications
Specifications
| Chemical Name or Material | 1-(4-Methanesulphonamidophenoxy)-2-[N-(4-methanesulphonamidophenethyl)-N-methylamino]ethane |
| CAS | 115256-11-6 |
| Assay Percent Range | >99% |
| Quantity | 25 mg |
| Product Line | Tocris |
| Molecular Formula | C19H27N3O5S2 |
| Formula Weight | Observed MW: 441.56 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?