missing translation for 'onlineSavingsMsg'
Learn More

Domiphen bromide, 97%

Product Code. 11317806
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317806 25 g 25g
11327806 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317806 Supplier Thermo Scientific Alfa Aesar Supplier No. L12930.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 538-71-6 | C22H40BrNO | 414.472 g/mol

Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-71-6
Molecular Formula C22H40BrNO
Molecular Weight (g/mol) 414.472
MDL Number MFCD00011769
InChI Key OJIYIVCMRYCWSE-UHFFFAOYSA-M
Synonym domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
PubChem CID 10866
IUPAC Name dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]

Specifications

Melting Point 117°C to 118°C
Quantity 25 g
Beilstein 3922742
Sensitivity Hygroscopic
Merck Index 14,3415
Solubility Information Soluble in ethanol,acetone,ethyl acetate,chloroform and water. Slightly soluble in benzene.
Formula Weight 414.48
Percent Purity 97%
Chemical Name or Material Domiphen bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.