missing translation for 'onlineSavingsMsg'
Learn More

Domiphen Bromide, TRC

Product Code. 30003444
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003444 1 g 1g
30007311 50 mg 50mg
30006619 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 30003444 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD5311051G
 Restricted Item
Estimated Shipment: 25-08-2026
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 538-71-6
Molecular Formula C22 H40 N O . Br
Molecular Weight (g/mol) 414.46
Synonym 1-Dodecanaminium, N,N-dimethyl-N-(2-phenoxyethyl)-, bromide, Ammonium, dodecyldimethyl(2-phenoxyethyl)-, bromide, Dodecyldimethyl(2-phenoxyethyl)ammonium bromide, (β-Phenoxyethyl)dimethyldodecylammonium bromide, Bradonit, Bradoral, Bradosol, Bradosol bromide, Domiphen bromide, Fungitex, Fungitex R, Modicare, N-Dodecyl-N-(2-phenoxyethyl)dimethylammonium bromide, NSC 39415, Neo-Bradoral, Oradol, PDDB, Phenododecinium bromide
IUPAC Name dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCC[N+](C)(C)CCOc1ccccc1

Tekniske data

Quantity 1 g
InChI Formula InChI=1S/C22H40NO.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-23(2,3)20-21-24-22-17-14-13-15-18-22;/h13-15,17-18H,4-12,16,19-21H2,1-3H3;1H/q+1;/p-1
Formula Weight 413.2293
Chemical Name or Material Domiphen Bromide
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.