missing translation for 'onlineSavingsMsg'
Learn More

Domiphen Bromide, TRC

Product Code. 30007311
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007311 50 mg 50mg
30003444 1 g 1g
30006619 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007311 Supplier Toronto Research Chemicals Supplier No. TRCD53110550MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-71-6
Molecular Formula C22 H40 N O . Br
Molecular Weight (g/mol) 414.46
Synonym 1-Dodecanaminium, N,N-dimethyl-N-(2-phenoxyethyl)-, bromide, Ammonium, dodecyldimethyl(2-phenoxyethyl)-, bromide, Dodecyldimethyl(2-phenoxyethyl)ammonium bromide, (β-Phenoxyethyl)dimethyldodecylammonium bromide, Bradonit, Bradoral, Bradosol, Bradosol bromide, Domiphen bromide, Fungitex, Fungitex R, Modicare, N-Dodecyl-N-(2-phenoxyethyl)dimethylammonium bromide, NSC 39415, Neo-Bradoral, Oradol, PDDB, Phenododecinium bromide
IUPAC Name dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
SMILES [Br-].CCCCCCCCCCCC[N+](C)(C)CCOc1ccccc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H40NO.BrH/c1-4-5-6-7-8-9-10-11-12-16-19-23(2,3)20-21-24-22-17-14-13-15-18-22;/h13-15,17-18H,4-12,16,19-21H2,1-3H3;1H/q+1;/p-1
Formula Weight 413.2293
Chemical Name or Material Domiphen Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.