missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Donepezil hydrochloride

Product Code. 18792704
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792704 50 mg 50mg
18762484 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792704 Supplier R&D Systems™ Supplier No. 4385/50
  • ONLINE EXCLUSIVE PRICE
    448.00 €
    423.15 € / 50mg
    Save 24.85 €
    5% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent AChE inhibitor

Potent acetylcholinesterase inhibitor (IC50 = 5.7 nM). Displays 1252-fold selectivity for AChE over BuChE (IC50 = 7138 nM for BuChE). Orally active.

Deuterated analog also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120011-70-3
Molecular Formula C24H30ClNO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 415.95
Purity >99%
Molecular Weight (g/mol) 415.96
Chemical Name or Material Donepezil hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.