missing translation for 'onlineSavingsMsg'
Learn More

Donepezil Hydrochloride, TRC

Product Code. 18032410
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032410 100 mg 100mg
18051230 250 mg 250mg
18021120 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18032410 Supplier Toronto Research Chemicals Supplier No. TRCD531750100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120011-70-3
Molecular Formula C24 H29 N O3 . Cl H
Molecular Weight (g/mol) 415.95
Synonym Donepezil hydrochloride, 1H-Inden-1-one, 2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, hydrochloride (1:1), 1H-Inden-1-one, 2,3-dihydro-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, hydrochloride (9CI), 1-Benzyl-4-[(5,6-dimethoxy-1-indanon-2-yl)methyl]piperidine hydrochloride, Aricept, Aricept D, Aricept ODT, BNAG, Donep, Dopezil, Dorent, E 2020, E 2020 (pharmaceutical)
IUPAC Name 2-[(1-benzylpiperidin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one;hydrochloride
SMILES Cl.COc1cc2CC(CC3CCN(Cc4ccccc4)CC3)C(=O)c2cc1OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H29NO3.ClH/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18;/h3-7,14-15,17,20H,8-13,16H2,1-2H3;1H
Formula Weight 415.1914
Percent Purity >95%
Chemical Name or Material Donepezil Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.