Learn More
Invitrogen™ Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP
Donkey Polyclonal Secondary Antibody
Brand:  Invitrogen™ PA186326
Product Code. 11513771
  • 956.00 € / 500µL
Description
Store at 4°C prior to reconstitution. After reconstitution store at -20°C. For long term storage the addition of 0.01% thiomersal is recommended. Reconstitute with 0.5 mL of distilled water. Donkey anti goat IgG antibody recognizes goat immunoglobulin G. This antibody has been absorbed against human, rabbit, rat, horse, guinea pig, hamster, chicken and mouse to minimize naturally occurring cross reactivities.
Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Goat IgG (H+L) Cross-Adsorbed
Polyclonal
HRP
Donkey
Affinity chromatography
RUO
Goat
Secondary Antibody
IgG
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 1.5% BSA and no preservative
Proprietary Immunogen.
500 μL
Secondary
4°C
Lyophilized