Learn More
Invitrogen™ Donkey anti-Rabbit IgG (H+L) ReadyProbes™ Secondary Antibody, Alexa Fluor™ 594
Donkey Polyclonal Secondary Antibody
Brand: Invitrogen™ R37119
Description
No pipetting or dilutions required. Just tip and drip two drops per mL buffer to stain your cells. This antibody shows minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat, and sheep serum proteins. Like other ReadyProbes reagents, AlexaFluor 594 Doneky anti-Rabbit IgG Antibody comes in a convenient dropper bottle.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Rabbit IgG (H+L) | |
| Polyclonal | |
| PBS with 0.1% BSA and <0.1% sodium azide | |
| Gamma Immunoglobin. | |
| 2 x 2.5 mL | |
| Secondary | |
| 4°C, store in dark | |
| Liquid | |
| Responsibly packaged |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Alexa Fluor 594 | |
| Donkey | |
| Affinity chromatography | |
| RUO | |
| Rabbit | |
| Secondary Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.