missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Donkey anti-Rabbit IgG (H+L) Secondary Antibody, Texas Red (Pre-adsorbed), Novus Biologicals™
Donkey Polyclonal Antibody
Brand: Novus Biologicals NB7609
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:1000 - 1:10000, Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Rabbit IgG whole molecule | |
| 0.2 mg | |
| Adaptive Immunity, Immunology | |
| This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat, and Sheep Serum Proteins. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat and Sheep Serum Proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Texas Red | |
| Donkey | |
| Multi-step | |
| RUO | |
| Secondary | |
| Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction