missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dopamine Competitive ELISA Kit

Product Code. 30824683
Click to view available options
Quantity:
96 Tests
This item is not returnable. View return policy

Product Code. 30824683

Brand: Invitrogen™ EEL144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Dopamine (C6H3(OH)2-CH2-CH2-NH2) is a catecholamine neurotransmitter in the brain. Its chemical name is 4-(2-aminoethyl)benzene-1,2-diol. Dopamine is a hormone released by the hypothalamus. Its main function is to inhibit the release of prolactin from the anterior lobe of the pituitary. It can be used as a sympathomimetic drug, producing effects such as increased heart rate and blood pressure.Detection of catecholamine-secreting cells.
TRUSTED_SUSTAINABILITY

Specifikationer

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Conjugate Unlabeled
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum, Tissue Homogenates
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <10
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL, Tissue Homogenate 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Dopamine
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 30 mins
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.