missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Dopamine hydrochloride

Product Code. 18702694
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702694 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702694 Supplier R&D Systems™ Supplier No. 3548/50
  • ONLINE EXCLUSIVE PRICE
    130.96 €
    116.55 € / 50mg
    Save 14.41 €
    11% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Endogenous agonist at dopamine D1-5 receptors

Dopamine hydrochloride is an endogenous neurotransmitter that acts as an agonist at dopamine D1-5 receptors. Synthesized in the substantia nigra and ventral tegmental area, and is a precursor in noradrenalin and adrenalin biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 62-31-7
Quantity 50 mg
Product Line Tocris
Molecular Formula C8H12ClNO2
Formula Weight 189.64
Purity >99%
Molecular Weight (g/mol) 189.64
Chemical Name or Material Dopamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.