missing translation for 'onlineSavingsMsg'
Learn More

Dopexamine Hydrochloride, TRC

Product Code. 30384684
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384684 5 mg
30382922 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384684 Supplier Toronto Research Chemicals Supplier No. TRCD5339005MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86484-91-5
Molecular Formula C22 H32 N2 O2 . 2 Cl H
Molecular Weight (g/mol) 429.42
Synonym Dopexamine dihydrochloride, 4-[2-[[6-[(2-Phenylethyl)amino]hexyl]amino]ethyl]benzene1,2-diol dihydrochloride, 1,2-Benzenediol, 4-[2-[[6-[(2-phenylethyl)amino]hexyl]amino]ethyl]-, hydrochloride (1:2), 1,2-Benzenediol, 4-[2-[[6-[(2-phenylethyl)amino]hexyl]amino]ethyl]-, dihydrochloride (9CI), Dopacard, Dopexamine hydrochloride, FPL 60278AR
IUPAC Name 4-[2-[6-(2-phenylethylamino)hexylamino]ethyl]benzene-1,2-diol;dihydrochloride
SMILES Cl.Cl.Oc1ccc(CCNCCCCCCNCCc2ccccc2)cc1O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H32N2O2.2ClH/c25-21-11-10-20(18-22(21)26)13-17-24-15-7-2-1-6-14-23-16-12-19-8-4-3-5-9-19;;/h3-5,8-11,18,23-26H,1-2,6-7,12-17H2;2*1H
Formula Weight 428.1997
Chemical Name or Material Dopexamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.