missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Dorsomorphin
Description
- Dorsomorphin inhibits the bone morphogenetic protein (BMP) pathway by targeting the type I BMP receptors activin receptor-like kinase (ALK) 2, ALK3, and ALK6
- It is also a potent inhibitor of AMP-activated protein kinase (AMPK; Ki = 109 nM) but does not significantly inhibit structurally related kinases such as ZAPK, SYK, PKCθ, PKA, or JAK3 (Bain et al.), Yu et al.)
- Promotes differentiation of neural progenitor cells from human pluripotent stem cells (Morizane et al.), Zhou et al.)
- Promotes differentiation of cardiomyocytes from mouse and human pluripotent stem cells (Hao et al.), Kattman et al.)
- Promotes differentiation of adipocytes and suppresses osteogenic differentiation of osteoblasts from human mesenchymal cells (Kim et al.)
Specifications
Specifications
| CAS | 866405-64-3 |
| Synonym | Compound C |
| Quantity | 10 mg |
| Molecular Formula | C24H25N5O |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?