missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Dorsomorphin dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Dorsomorphin dihydrochloride is a potent inhibitor of AMP-activated protein kinase (AMPK) (Ki = 109 nM). Displays no significant activity on several structurally related kinases including ZAPK, SYK, PKCθ, PKA and JAK3. Inhibits AMPK activation induced by AICAR (Cat. No. 2840) and Metformin (Cat. No. 2864). Dorsomorphin dihydrochloride also inhibits bone morphogenetic protein (BMP) type I receptors (ALK2, ALK3 and ALK6), promotes cardiomyogenesis in mouse embryonic stem cells (ESCs) in vitro and promotes neural differentiation of hPSCs as part of a chemical cocktail. Shown to induce autophagy in cancer cell lines via a mechanism independent of AMPK inhibition.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 472.41 |
| Purity | >98% |
| Molecular Weight (g/mol) | 472.414 |
| Chemical Name or Material | Dorsomorphin dihydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction