missing translation for 'onlineSavingsMsg'
Learn More

Dorsomorphin, MedChemExpress

Product Code. 16471837
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471837 50 mg 50mg
16458367 5 mg 5mg
16416186 100 mg 100mg
16405678 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471837 Supplier Medchem Express Supplier No. HY13418A
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor (Ki=109 nM in the absence of AMP). Dorsomorphin (BML-275) selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Dorsomorphin induces autophagy.

Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor (Ki=109 nM in the absence of AMP). Dorsomorphin (BML-275) selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Dorsomorphin induces autophagy.

Specifications

Chemical Name or Material Dorsomorphin
CAS 866405-64-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H25N5O
Quantity 50 mg
Synonym Compound C BML-275
Solubility Information 1M HCl : 50 mg/mL (125.16 mM; ultrasonic and adjust pH to 1 with HCl) ∣DMSO : 5 mg/mL (12.52 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : 3.33 mg/mL (8.34 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C12=C(C3=CC=NC=C3)C=NN1C=C(C4=CC=C(OCCN5CCCCC5)C=C4)C=N2
Molecular Weight (g/mol) 399.49
Formula Weight 399.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.