missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KARTELL DOSILAB
Description
- Completely autoclavable at +121°C without need for disassembly.
- PTFE piston with silicone O-ring ensuring a high degree of accuracy.
- Internal cylinder in borosilicate glass protected by PP sheath.
- Valve housing: PTFE body in a single piece ensuring high chemical resistance.
- Accuracy and Reproducibility: according to the ISO 8655 standards.
- Compatibility: excellent with all reagents including organic solvents and concentrated acids, except hydrofluoric acid (HF).
- Recalibration: simple and quick with its special accessory.
- All the internal components that during use are in contact with liquid solutions are made of PTFE, FEP and borosilicate glass. They therefore possess excellent compatibility with all reagents (even organic solvents and concentrated acids), except for hydrofluoric acid.
Specifications
Specifications
| Accuracy | ± 0.050 mL |
| Product Type | Adjustable Volume Dispenser |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 8655 |
| Color | White |
| Channels | 1 |
| Material | FEP, PTFE, Borosilicate Glass, Polypropylene |
| Volume (Metric) | 10 mL |
| Display | Analog |
| Increments | 0.2 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?