missing translation for 'onlineSavingsMsg'
Learn More

KARTELL DOSIPET B

Product Code. 10798682
Change view
Click to view available options
Volume (Metric):
2.5 mL
5 mL
10 mL
100 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10798682 100 mL Each
10335971 2.5 mL Each
10172711 5 mL Each
10011031 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10798682 Supplier KARTELL Supplier No. 8306000
  • ONLINE EXCLUSIVE PRICE
    915.00 €
    862.92 € / Pack of 1
    Save 52.08 €
    5% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Dosipet B is a series of adjustable volume repetitive dosing devices. They are made of high quality materials ensuring considerable mechanical and chemical resistance.

  • For universal use; ideal for use with reagent of average aggressiveness.
  • It can be used with every water solution type (except those containing hydrofluoric acid), marked by a high degree of convenience and reliability. It is equipped with a 3.3 borosilicate glass cylinder and PTFE piston.
  • Precise and quick volume adjustment using the analogue slider
  • Precise graduated scale to ensure reproducibility of the dispensed volumes
  • No loss of reagent when eliminating air or filling
  • Dispensing without bubbles forming
  • The piston completely transfers the liquid with every movement
  • Total recovery of the liquid in the discharge tube by turning the delivery tip 180°
  • Autoclavable at +121°C for 20 minutes

Specifications

Accuracy ± 0.6 mL
Product Type Dosispet B
Autoclavable Autoclavable
Certifications/Compliance None
Color White
Channels 1
Material Borosilicate Glass, PTFE
Volume (Metric) 100 mL
Display Analog
Increments 2.0 mL
Volume (English) 3.38 oz.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.