missing translation for 'onlineSavingsMsg'
Learn More

Dothiepin, TRC

Product Code. 30137844
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137844 5 mg 5mg
30136395 10 mg 10mg
17913275 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137844 Supplier Toronto Research Chemicals Supplier No. TRCD5365005MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-53-1
Molecular Formula C19H21NS
Molecular Weight (g/mol) 295.44
Synonym 1-Propanamine, 3-dibenzo[b,e]thiepin-11(6H)-ylidene-N,N-dimethyl-, Dibenzo[b,e]thiepin-δ11(6H),γ-propylamine, N,N-dimethyl- (7CI,8CI), 3-Dibenzo[b,e]thiepin-11(6H)-ylidene-N,N-dimethyl-1-propanamine, Dibenzo[b,e]thiepin, 1-propanamine deriv., Dosulepin, Dosulepine, Dothep, Dothiepin, IZ 914, Prothiaden, Prothiadene
SMILES CN(C)CC\C=C\1/c2ccccc2CSc3ccccc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H21NS/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19/h3-6,8-12H,7,13-14H2,1-2H3/b17-11-
Formula Weight 295.1395
Percent Purity >95%
Chemical Name or Material Dothiepin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.