missing translation for 'onlineSavingsMsg'
Learn More

Water, Double Distilled, Pyrogen-Free, Chem-Lab

Product Code. 11963345
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
10 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11963345 10 L 10 L
11983084 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11963345 Supplier Chem-Lab Supplier No. CL02.0208.9010
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for laboratory use. Water, Double Distilled, Pyrogen-Free, Chem-Lab has been ultra-purified to remove bacteria, organic and inorganic particles, viruses, minerals, salts and metals, at LF<5μS. Packaged in a sterilized container to maintain purity.

Distilled water is produced by boiling water and then condensing the vapor into a clean container, leaving solid contaminants behind. Double-distilled water produces an even higher level of purification. It is prepared by slow boiling the uncontaminated condensed water vapor from a prior slow boiling.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.016g/mol
Filtered Through LF < 5μS
Treatment(s) Double-Distilled
Density 1.00g/mL
Quantity 10 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.