missing translation for 'onlineSavingsMsg'
Learn More

Kolmi Hopen™ Double Elastic Polypropylene Clip Caps in Wall Dispenser

Product Code. 17790129 Shop All Medicom Products
Change view
Click to view available options
Color:
Black
Blue
Green
Unit Size:
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17790129 Black Pack of 250
17700139 Blue Pack of 250
17710139 Green Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17790129 Supplier Kolmi Hopen™ Supplier No. 70.111BLACK
  • ONLINE EXCLUSIVE PRICE
    11.72 €
    7.87 € / Pack of 250
    Save 3.85 €
    32% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These double elastic clip caps come in wall-mounted dispensers for convenient use.

  • Covers the full head, as required by the HACCP process
  • Extra comfort with double elastic to ensure excellent retention
  • Ability to use color code for tracking
  • 12 g/m2 fabric weight

Specifications

Material Polypropylene
Color Black
Disposable Single-use
Design Type Clip Cap
Sterility Non-sterile
Certifications/Compliance EN ISO 9001
Quantity 250 Pk.
Dimensions (L x W) 40 x 55 cm
Includes Caps and Wall Dispenser
For Use With (Application) Industrial, Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.