missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Double Pore Knick Electrode

Product Code. 12983187
Change view
Click to view available options
Length (Metric):
35 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
12983187 35 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12983187 Supplier Hamilton™ Supplier No. 242066
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Accuracy (mV) 0 ± 20mV
Accuracy (Temperature) 0°C to 60°C
Diameter (Metric) 12 mm
Electrical Requirements 0 ± 20 mV, 57 to 59 mV
Length (Metric) 35 mm
Material PEEK
pH Range 0 to 14
Temperature (Metric) Range 0°C to 60°C
Certifications/Compliance Yes, with measured values in Buffer pH 4.01 and pH 7.00
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.