Learn More
Schneider Gerd™ Double Spatula
Use this spatula for a variety of pharmaceutical applications. Schneider Gerd™ Double Spatulas are ideal for use with a wide range of compounds.
5.25 €
Products 1
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
10001001
View Documents Promotion Details
Schneider Gerd™
8301
130 mm
5.25 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Spatulas are ideal for working with powders, stirring and mixing
  • Two end shapes offer increased versatility
  • Perfect for miniscule amounts of chemicals
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant