missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Globe Scientific Double Tab Snap Caps for Blood Drawing Tubes, Glass and Plastic Tubes
Shop All Globe Scientific ProductsDescription
- Precision molded from LDPE
- Comes with two thumb tabs for easy removal of cap
- Provides tight seal during centrifugation, agitation and refrigeration procedures
- Protects samples from cross-contamination and highly infectious aerosols
- Color-coded for easy sample identification
- Color: green
Specifications
Specifications
| Includes | 2 thumb tabs |
| Product Type | Snap Cap |
| Material | LDPE |
| Color | Green |
| Sterility | Non-sterile |
| For Use With (Equipment) | 13 mm Tubes |
| Disposable | Single-use |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?