Learn More
Gilac™ Double-wall Container with Stainless Steel Wheel Housing
Product Code. 17948781
Change view
Click to view available options
Capacity (Metric):
310 L
500 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17948781 310 L Each
17938821 500 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17948781 Supplier Gilac™ Supplier No. G156921
Only null left
Add to basket
Add to basket
Double-wall Container with Stainless Steel Wheel Housing is ideal for transporting crushed ice, prepared food and marinated items. Wide range of uses.
  • Ideal for transporting crushed ice, prepared food and marinated items.
  • Wide range of uses.
  • No gaps nor overlaps and no sharp corners.
  • Double-walled offering excellent thermal insulation and excellent mechanical resistance.
  • Container comprising a drainage outlet with drain plug.
  • Rotational molded container with 160 mm, polypropylene wheels with synthetic rubber tires (2 fixed and 2 pivoting) and stainless steel wheel housing.
  • Stainless steel clevis recommended in humid environments (no damage to the bearings even in the presence of water).
  • Can be stored on 800 x 1200 pallets, stackable upside down.
  • Integral high-flow drainage.
  • 1/4 turn angle stop valve optional extra.
  • Volume in litres is equivalent to the container filled to the brim.
Specifications
Type Storage Tote
Material HDPE
Color White
Shape Rectangular
Capacity (English) 81.9 gal.
Capacity (Metric) 310 L