missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 1X8, 50-100 mesh, ion-exchange resin

Product Code. p-2885571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10469900 100 g Plastic bottle 100g
10276350 500 g Plastic bottle 500g
13408468 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10469900 Supplier Thermo Scientific Acros Supplier No. 202991000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Dowex 1X8, C29H34ClN, CAS Number-12627-85-9, dowex 1x8 50-100 cl, dowex 1x2 50-100 cl, 500g, White, 01,511; 04,267; 06,302; 07,182; 08,263, BBQMUEOYPPPODD-UHFFFAOYSA-M, 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride, MFCD00132718, 432.048, Plastic bottle | CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
SMILES *

Specifications

Physical Form Powder and/or Chunks
Particle Size 15 % max. (on 35 mesh, cumulative) 15 % max. (through 80 mesh)
Packaging Plastic bottle
Volume Total Capacity wet volume capacity, Cl form (meq/mL):> 1.2
Water Retention Capacity Cl form: 43 to 48%
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Color White
Quantity 100 g
IUPAC Name 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
Chemical Name or Material Dowex™ 1X8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.