missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 50WX4 100-200 (H)

Product Code. 11318815
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318815 50 g 50g
11328815 250 g 250g
11338815 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11318815 Supplier Thermo Scientific Alfa Aesar Supplier No. L13917.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 11113-61-4

Dowex™ 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11113-61-4
MDL Number MFCD00132725

Specifications

Quantity 50 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX4 100-200 (H)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.