missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX8 200-400 (H)

Product Code. 11328825
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328825 1000 g 1kg
11308825 50 g 50g
11318825 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11328825 Supplier Thermo Scientific Alfa Aesar Supplier No. L13922.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 11119-67-8

AmberChrome™ 50WX8 Ion Exchange Resin) is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
AmberChrome™ 50WX8 Ion Exchange Resin is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 50WX8 200-400 (H)
compliance-icons
Product Identifier
  • Dowex 50WX8 200-400 (H)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dowex 50WX8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.