missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 50WX8, 200-400 mesh, ion-exchange resin

Product Code. 10454415 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10454415

Brand: Thermo Scientific Acros 335350025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 11119-67-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11119-67-8
MDL Number MFCD00132726
IUPAC Name AmberChrom™ 50WX8 Ion Exchange Resin

Specifications

Physical Form Beads
Particle Size 15% max. (on 100 mesh, cumulative), 15% max. (through 325 mesh)
Packaging Plastic bottle
Volume Total Capacity wet volume capacity, H form min. (meq/mL): 1.7
Water Retention Capacity H form: 50 to 58%
Color Beige to Brown
Quantity 2.5 kg
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Dowex™ 50WX8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.