missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 50WX2, 50-100 mesh, ion-exchange resin

Product Code. 10286400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
2.5kg
500g
Product Code. Quantity Packaging unitSize
10286400 100 g Plastic Bottle 100g
10296400 500 g Plastic Bottle 500g
10287113 2.5 kg Plastic Bottle 2.5kg
3 options
This item is not returnable. View return policy

Product Code. 10286400

Brand: Thermo Scientific Acros 203021000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12612-37-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12612-37-2
MDL Number MFCD00132722

Specifications

Physical Form Beads
Particle Size Particle size analysis: on 20 mesh (cumulative): <15%, through 70 mesh: <15%
Packaging Plastic Bottle
Volume Total Capacity wet volume capacity, H form (meq/mL):> 0.6
Water Retention Capacity H form: 74 to 82%
Fieser 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,511; 04,266; 05,355; 06,302; 09,256; 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176
Quantity 100 g
Chemical Name or Material Dowex™ 50WX2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.