missing translation for 'onlineSavingsMsg'
Learn More

Dowex Marathon C, H+ form, Thermo Scientific™

Product Code. p-7028414
Change view
Click to view available options
Quantity:
250 g
1000 g
2500 g
Unit Size:
1000g
2500g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451110 250 g 250g
11431110 1000 g 1000g
11441110 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451110 Supplier Thermo Scientific Alfa Aesar Supplier No. B25721.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dowex Marathon C
Name Note H+ form
CAS 150604-77-6
MDL Number MFCD00212649
Density 1.2
Quantity 250 g
Solubility Information Insoluble in water.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.