missing translation for 'onlineSavingsMsg'
Learn More

Dowtherm A, TRC

Product Code. 30370554
Change view
Click to view available options
Quantity:
250 mL
missing translation for 'unitSize':
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30370554 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30370554 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD537100250ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dowtherm A
CAS 8004-13-5
Molecular Formula C12H10O.C12H10
Quantity 250 mL
Synonym 1,1'-Biphenyl Mixture With 1,1'-Oxybis[benzene], Biphenyl-diphenyl Ether Mixture, Biphenyl-phenyl Ether Mixture, Dinil, Dinyl, Diphyl, Dowtherm, Therminol VP, Therminol VP 1
InChI Formula InChI=1S/C12H10O.C12H10/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H;1-10H
SMILES C1(OC2=CC=CC=C2)=CC=CC=C1.C3(C4=CC=CC=C4)=CC=CC=C3
IUPAC Name oxydibenzene 1,1'-biphenyl
Molecular Weight (g/mol) 324.42
Formula Weight 324.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.