missing translation for 'onlineSavingsMsg'
Learn More

Doxepin-d3 Hydrochloride, TRC

Product Code. 30135231
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135231 1 mg 1mg
30137690 25 mg 25mg
17909309 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30135231 Supplier Toronto Research Chemicals Supplier No. TRCD5500021MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 347840-07-7
Molecular Formula C19 2H3 H18 N O . Cl H
Molecular Weight (g/mol) 318.86
Synonym Doxepin-D3 Hydrochloride (cis/trans), 3-Dibenz[b,e]oxepin-11(6H)-ylidene-N-methyl-N-(methyl-d3)-1-propanamine hydrochloride
IUPAC Name 3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N-methyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride
SMILES Cl.[2H]C([2H])([2H])N(C)CCC=C1c2ccccc2COc3ccccc13

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/i1D3;
Formula Weight 318.1578
Chemical Name or Material Doxepin-d3 Hydrochloride (cis/trans)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.