missing translation for 'onlineSavingsMsg'
Learn More

Doxepin Hydrochloride, TRC

Product Code. 30137552
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137552 2.5 g 2.5g
30133862 5 g 5g
18000820 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137552 Supplier Toronto Research Chemicals Supplier No. TRCD5500002.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229-29-4
Molecular Formula C19 H21 N O . Cl H
Molecular Weight (g/mol) 315.84
Synonym Doxepin Hydrochloride, 3-Dibenz[b,e]oxepin-11(6H)-ylidene-N,N-dimethyl-1-propanamine hydrochloride, Dibenz[b,e]oxepin, 1-propanamine deriv., Adapin, Aponal, Curatin, Doxepin hydrochloride, Doxepine hydrochloride, N,N-Dimethyldibenz[b,e]oxepin-δ11(6H),γ-propylamine hydrochloride, P 3693A, Quitaxon, Sinequan, Sinequin
IUPAC Name (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CC\C=C\1/c2ccccc2COc3ccccc13

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11+;
Formula Weight 315.139
Chemical Name or Material Doxepin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.