missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Doxycycline hyclate, 98%

Product Code. 15510554 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15510554 25 g 25g
15500554 5 g 5g
15520554 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15510554 Supplier Thermo Scientific Chemicals Supplier No. 446060250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Doxycycline hyclate, Doryx, CAS # 24390-14-5, is the hyclate salt form of the broad-spectrum antibiotic known as doxycycline.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Doxycycline hyclate is a broad-spectrum bacteriostatic antibiotic of the tetracycline class
  • Doxycycline inhibits bacterial growth by preventing polypeptide formation via inhibition of at least one ribosomal subunit
  • Known to reversibly bind the 30S ribosomal subunit and inhibit formation of the aminoacyl-tRNA/mRNA-ribosome complex
  • Inhibits matrix metalloproteinases in many microorganisms

Applications

  • Effective against the bacteria that cause Lyme disease, chlamydia, rickettsia and mycoplasma; suitable for antibiotic resistance testing and study of cellular mechanisms
  • Intended for laboratory use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24390-14-5
Molecular Formula 0·5 C2H6O
Molecular Weight (g/mol) 512.94
InChI Key ISZOFWGVNMFENH-YDLUHMIOSA-N
Synonym doryx, doxycycline hyclate, doxycycline, hydrochloride, hemiethanolate, hemihydrate;
PubChem CID 131676157
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
SMILES CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Merck Index 15,3488
Solubility Information Solubility in water: soluble.
Formula Weight 512.94
Percent Purity 98%
Chemical Name or Material Doxycycline hyclate
compliance-icons compliance-icons
Product Identifier
  • Doxycycline hyclate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.