missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ DPD 1 Buffer Solution

Product Code. 10722203
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10722203 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10722203 Supplier Lovibond™ Supplier No. 471010
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For photometric determination of free/total chlorine in water testing. Lovibond™ Tintometer™ DPD 1 Buffer Solution is an environmentally friendly reagent packaged in a color-coded blue dropper bottle for precise liquid dosing.

For photometric determination of free/total chlorine in water testing. Lovibond™ Tintometer™ DPD 1 Buffer Solution is an environmentally friendly reagent packaged in a color-coded blue dropper bottle for precise liquid dosing.

  • Color coded bottle for quick visual identification of the reagent
  • Convenient dropper bottle provides precise dosing of liquid reagents
  • Environmentally friendly formulation free of boric acid
  • For use with Lovibond™ photometers
  • Carefully developed formula for accurate analysis results

Specifications

Analytes Chlorine
Color Colorless
pH Range 7 at 20°C
Format Aqueous Solution
For Use With (Application) Water sample analysis (e.g. chlorine determination)
For Use With (Equipment) Lovibond™ photometer
Packaging Blue Dropper Bottle
Quantity 15 mL
Range 0.02 to 4mg/L total chlorine
Sample Size 10 mL
Storage Requirements Room temperature unopened, Refrigerated after
Type Buffer solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.