missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DPO-1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Blocker of IKur ultrarapid delayed rectifier potassium current and KV1.5 channels (IC50 = 0.31 μM for rKV1.5). Displays selectivity for inhibition of IKur over Ito (8-fold), IK1, IKr and IKs (20-fold) in native myocytes and selectivity for rat recombinant KV1.5 over KV3.1 (∽ 15-fold). Increases action potential duration in atrial but not ventricular myocytes and prevents atrial arrhythmia.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 340.44 |
| Purity | >99% |
| Molecular Weight (g/mol) | 340.45 |
| Chemical Name or Material | DPO-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction