missing translation for 'onlineSavingsMsg'
Learn More

DPPC, MedChemExpress

Product Code. 16484298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484298 10 mg 10mg
16480627 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484298 Supplier Medchem Express Supplier No. HY109506
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.

DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.

Specifications

Chemical Name or Material DPPC
CAS 63-89-8
Color White
Purity Grade Notes Research
Molecular Formula C40H80NO8P
Quantity 10 mg
Synonym 129Y83
Solubility Information Ethanol : 25 mg/mL (34.06 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (6.81 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CCCCCCCCCCCCCCCC(O[C@H](COC(CCCCCCCCCCCCCCC)=O)COP(OCC[N+](C)(C)C)([O-])=O)=O
Molecular Weight (g/mol) 734.04
Formula Weight 734.04
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.