missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Butadiene (OV) XXS Replacement Sensor

Product Code. 12607316
Change view
Click to view available options
Components Measured:
Butadiene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
12607316 Butadiene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12607316 Supplier Dräger™ Supplier No. 6811530
  • ONLINE EXCLUSIVE PRICE
    1139.55 €
    445.78 € / Pack of 1
    Save 693.77 €
    60% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

XXS Replacement Sensor, for use with Pac 7000 and X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 100 ppm
  • Resolution: 1 ppm
  • Butadiene (OV)

Specifications

Components Measured Butadiene
For Use With (Equipment) Pac 7000, X-am 5000 Multigas Detectors
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.