missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Carbon Monoxide/Hydrogen Sulfide (CO/H2S) XXS Replacement Sensor

Product Code. 13467346
Change view
Click to view available options
Components Measured:
Carbon Monoxide, Hydrogen Sulfide
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
13467346 Carbon Monoxide, Hydrogen Sulfide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13467346 Supplier Dräger™ Supplier No. 6811410
  • ONLINE EXCLUSIVE PRICE
    607.05 €
    443.41 € / Pack of 1
    Save 163.64 €
    26% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

XXS Replacement Sensor, for use with X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 2000 ppm (CO); 0-200 ppm (H2S)
  • Resolution: 2 ppm (CO); 1 ppm (H2S)
  • Carbon Monoxide (CO); Hydrogen Sulfide (H2S)

Specifications

Components Measured Carbon Monoxide, Hydrogen Sulfide
For Use With (Equipment) X-am 5000 Multigas Detector
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.