Learn More
Dräger™ CMS Chip Measurement System
Product Code. 12394649
Change view
Click to view available options
Type:
CMS Analyzer
CMS Remote System
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12394649 CMS Remote System Each
10629932 CMS Analyzer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12394649 Supplier Dräger™ Supplier No. 6405060
  • ONLINE EXCLUSIVE PRICE
    676.28 €
    498.85 € / Pack of 1
    Save 177.43 €
    26% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
Comes with carrying case and batteries. Optional remote system (sold separately) includes an additional pump, a screwdriver, and a 9 ft. (3m) extension hose to sample contaminants in inaccessible areas.
Requires
CMS chips (sold separately)
Uses an analyzer and substance-specific chips to obtain accurate, reliable measurements of gas and vapor concentrations
Bar code on each chip contains specific information such as gas type, measurement range, and other necessary parameters for measurement evaluation; when a chip is inserted into the analyzer, the bar code is read and interpreted. Choose from more than 50 interchangeable chips (sold separately).
  • Mass-flow-regulated pump system draws a constant flow of sample through the chip, compensating for any pressure fluctuation
  • Easy to use, with simple menu commands and one-switch operation
  • Chip contains 10 measurement channels filled with a substance-specific reagent that provides a quick reaction with the contaminant
  • Intelligent interaction ensures high level of accuracy
  • Data recorder gives contaminant, concentration, date, time, and location; stores up to 50 measurements
Order Info
Compact, rugged housing measures approximately 1.5L x 3.5W x 8 in. H (3.8 x 8.9 x 20.3cm). Net weight: 1.63 lb. (730g). Uses four AA alkaline batteries (included).
Certifications
Intrinsically safe and certified according to UL, MSHA and CENELEC standards. (UL and CSA Classified as Intrinsically Safe for use in Class I, Div. 1, Groups A-D, T4.)
Specifications
Type CMS Remote System