Learn More
Dräger™ CMS Chips: Hydrogen Cyanide
Product Code. 10719772
Change view
Click to view available options
Measurement Range:
2 to 50
Unit Size:
Each
Product Code. Measurement Range Unit Size
10719772 2 to 50 Each
1 options
Product Code. 10719772 Supplier Dräger™ Supplier No. 6406100
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrogen Cyanide CMS Chip
Humidity Range 1 to 20mg/L
Measurement Range 2 to 50
Response Time 30 to 260 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 30°C