Learn More
Dräger™ CMS Chips: Hydrogen Peroxide
Product Code. 12364619
Change view
Click to view available options
Measurement Range:
0.2 to 2
Unit Size:
Each
Product Code. Measurement Range Unit Size
12364619 0.2 to 2 Each
1 options
Product Code. 12364619 Supplier Dräger™ Supplier No. 6406440
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrogen Peroxide CMS Chip
Humidity Range 1 to 20mg/L
Measurement Range 0.2 to 2
Response Time 180 to 600 sec.
Temperature Range (English) 50°F to 86°F
Temperature Range (Metric) 10°C to 30°C