Learn More
Dräger™ CMS Chips: Hydrochloric Acid
Product Code. 10259801
Change view
Click to view available options
Measurement Range:
1 to 25
20 to 500
Response Time:
15 to 110 sec.
6 to 80 sec.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time unitSize
10259801 1 to 25 15 to 110 sec. Each
10513432 20 to 500 6 to 80 sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10259801 Supplier Dräger™ Supplier No. 6406090
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrochloric Acid CMS Chip
Humidity Range 1 to 10mg/L
Measurement Range 1 to 25
Response Time 15 to 110 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C