Learn More
Dräger™ CMS Chips: Ozone
Product Code. 10166004
Change view
Click to view available options
Measurement Range:
25 to 1000
Unit Size:
Each
Product Code. Measurement Range Unit Size
10166004 25 to 1000 Each
1 options
Product Code. 10166004 Supplier Dräger™ Supplier No. 6406430
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Ozone CMS Chip
Humidity Range 1 to 25mg/L
Measurement Range 25 to 1000
Response Time 100 to 600 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C