Learn More
Dräger™ CMS Chips: Phosphine
Product Code. 12344639
Change view
Click to view available options
Measurement Range:
1 to 25
20 to 500
200 to 5000
Response Time:
20 to 200 sec.
25 to 220 sec.
50 to 600 sec.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time unitSize
12344639 1 to 25 50 to 600 sec. Each
12354639 20 to 500 25 to 220 sec. Each
12364639 200 to 5000 20 to 200 sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12344639 Supplier Dräger™ Supplier No. 6406410
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
Suitable for 10 measurements of one gas.
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Phosphine CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 1 to 25
Response Time 50 to 600 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C