Learn More
Dräger™ CMS Chips: Vinyl Chloride
Product Code. 12354649
Change view
Click to view available options
Measurement Range:
0.3 to 10
10 to 250
Response Time:
30 to 420 sec.
45 to 100 sec.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time Unit Size
12354649 10 to 250 45 to 100 sec. Each
12344649 0.3 to 10 30 to 420 sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12354649 Supplier Dräger™ Supplier No. 6406230
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Vinyl Chloride CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 10 to 250
Response Time 45 to 100 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C