missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Carbon Dioxide (CO2) XXS Replacement Sensor

Product Code. 15348606
Change view
Click to view available options
Components Measured:
Carbon Dioxide
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
15348606 Carbon Dioxide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15348606 Supplier Dräger™ Supplier No. 6810889
  • ONLINE EXCLUSIVE PRICE
    575.10 €
    421.88 € / Pack of 1
    Save 153.22 €
    26% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

XXS Replacement Sensor, for use with Pac 7000, X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 5% vol.
  • Resolution: 0.1% vol.
  • Carbon Dioxide (CO2)

Specifications

Components Measured Carbon Dioxide
For Use With (Equipment) Pac 7000, X-am 5000 Multigas Detectors
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.