missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Pac™ III Sensors

Product Code. 15155187
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide (XS)
Oxygen (XS)
Measurement Range:
0 to 100
0 to 25%
Warranty:
3 Years
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Measurement Range Warranty Unit Size
15155187 Hydrogen Sulfide (XS) 0 to 100 3 Years Each
11362080 Oxygen (XS) 0 to 25% 3 Years Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15155187 Supplier Dräger™ Supplier No. 6809110
  • ONLINE EXCLUSIVE PRICE
    514.40 €
    382.60 € / Pack of 1
    Save 131.80 €
    25% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provide fast and accurate responses to hazardous gas levels

  • For use with Pac™ III single gas monitors

Specifications

Components Measured Hydrogen Sulfide (XS)
Humidity Range 10 to 90%
Measurement Range 0 to 100
Pressure Range 700 to 1300mbar
Resolution 1
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 3 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.