missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ Short-Term Detector Tubes

Product Code. 10454292
Change view
Click to view available options
Measurement Range:
0.2 to 2%
40 to 800
Unit Size:
5 tests
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10454292 0.2 to 2% Pack of 10
10707674 40 to 800 5 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10454292 Supplier Dräger™ Supplier No. 8101511
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Glass detector tubes for the rapid, precise measurement of hydrogen gas concentrations using a gas detection pump.

DrägerTubes are glass vials containing a chemical reagent designed to react with specific chemicals or gas families. When a calibrated 100 mL air sample is drawn through the tube using a Dräger Accuro™ bellows pump, the presence of the target analyte causes a color change, with the length of the stain indicating the measured concentration.

  • Application: Detection of gas concentrations.
  • Standard Range of Measurement: 0.2 Vol% [at 20°C, 1013 hPa].
  • Measurement Time: 1 min.
  • Usage: Suitable for manual operation with Dräger Accuro pumps or automated operation with Accuro 2000 or Quantimeter 1000.
  • Field Applications: Concentration peak determination, exposure measurement, leak detection, and analysis in confined spaces such as sewers, shafts, or tanks.

Specifications

Components Measured Hydrogen
For Use With (Equipment) Accuro
Type Gas Detection Tube
Measurement Range 0.2 to 2%
Pressure Range 1013hpa
Temperature Range (English) 68°F to 104°F
Temperature Range (Metric) 20° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.